Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system. LC Use With Instru 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system. LC Use With Instru 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II Purification system 1290 Infinity II Purification system. LC Use With Instru 1 * 1 Pce HEWLG1314-60025 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou