Specifications:- Inner Diameter (ID): 2 mm. Packed Column Length: 4 ft (1.22 m). Packed Column Material: UltiMetal. Packed Column Me sh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: HayeSep N. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 2 mm. Packed Column Length: 4 ft (1.22 m). Packed Column Material: UltiMetal. Packed Column Me sh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: HayeSep N. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 2 mm. Packed Column Length: 4 ft (1.22 m). Packed Column Material: UltiMetal. Packed Column Me sh: 80/100. Packed Column Outer Diameter (OD): 0.125 in. Packed Column Support: HayeSep N. 1 * 1 Pce HEWLG3591-81184 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou