Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Purification system. LC Use With Instrument: G711 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Purification system. LC Use With Instrument: G711 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Detector. LC Supplies Brand: InfinityLab. LC System Compatibility: 1260 Infinity II LC System 1260 Infinity II Purification system. LC Use With Instrument: G711 1 * 1 Pce HEWLG7115-60001 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou