Specifications:- LC Application Compatibility: Bio-compatible. LC Component Type: Loop. LC System Compatibility: 1290 infinity II Bi o LC System. LC Use With Instrument: G7137A. Sample Loop Material: MP35N. Volume: 40 uL. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Bio-compatible. LC Component Type: Loop. LC System Compatibility: 1290 infinity II Bi o LC System. LC Use With Instrument: G7137A. Sample Loop Material: MP35N. Volume: 40 uL. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Bio-compatible. LC Component Type: Loop. LC System Compatibility: 1290 infinity II Bi o LC System. LC Use With Instrument: G7137A. Sample Loop Material: MP35N. Volume: 40 uL. 1 * 1 Pce HEWLG7137-60400 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou