Homogénéiseurs de paillasse, JumboMix® 3500
Fournisseur: INTERSCIENCE
031230 032230 033230
INSC031230EA 0 EUR InStock
INSC031230 INSC032230 INSC033230
Homogénéiseurs de paillasse, JumboMix® 3500
Homogénéisateurs Homogénéisateurs de laboratoire
JumboMix® 3500 are highly efficient lab blenders for solid sample preparation. With a sterile bag, they guarantee optimal bacterial extraction in less than 60 seconds and eliminate all risks of cross contamination. The JumboMix® 3500 models enable fast blending of large samples (up to 400g) for numerous applications in microbiology.
  • Door opens at 270° and paddles stop side by side at end of blending for easy extraction of bags
  • Paddles are adjustable which together with the stainless steel construction make cleaning easy
  • Multifunctional digital screen
  • Simple control of blending time, power and speed
  • Variable speed and sample range of 200 to 3500 ml
  • Q-Tight® closing system prevents risk of liquid leaking from the top of the bag
Information de livraison: All models supplied with user manual and free pack of sterile filter bags. The JumboMix ®3500 VW and WarmMix® models are supplied with a security drip tray.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité