Synonyms:
(2S,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol
J60731.MA
J60731.MAEA
533
EUR
InStock
J60731.MA
(-)-Gallocatechin
(-)-Gallocatechin
CAS No.: 3371-27-5
Molecular Formula: C15H14O7
Formula Weight: 306.27
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD01632616
Applications: Polyphenolic compound from green tea. Possesses free-radical scavenging ability