Synonyms:
3,4-Dihydroxy-N-[3-(4-hydroxyphenyl)-1-methylpropyl]-beta-phenethylamine Hydrochloride
J61333.03
J61333.03EA
303
EUR
InStock
J61333.03
Dobutamine hydrochloride
Dobutamine hydrochloride
CAS No.: 49745-95-1
Molecular Formula: C18H23NO3.HCl
Formula Weight: 337.85
Physical Form: Powder
Appearance: Off-white to white
Melting Point: 184-186°
Solubility: Soluble in water or ethanol with warming. Also soluble in methanol
MDL No.: MFCD00153795
Applications: A beta-1 adrenoceptor agonist with weak beta-2 adrenoceptor activity and some action at the alpha-1 adrenoceptor