J61354.MF
J61354.MFEA
253
EUR
InStock
J61354.MF
N''-Hydroxyguanidine acetate
N''-Hydroxyguanidine acetate
CAS No.: 6345-29-5
Molecular Formula: CH5N3O.0.5H2SO4
Formula Weight: 124.11
Storage Temperature: -30°C to -10°C
Physical Form: Crystals
Appearance: White
Melting Point: 132-134° dec.
MDL No.: MFCD00042033
Applications: Reacts with nitric oxide forming an adduct which is a potent and stable vasodilator