J62213.AK
J62213.AKEA
58.8
EUR
InStock
J62213.AK
Polyethylene glycol-lithium acetate solution
Polyethylene glycol-lithium acetate
Common solution for protein and DNA isolation from yeast
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless
Common solution for protein and DNA isolation from yeast