Synonyms:
6-Chloroguanineriboside, (2R,3R,4S,5R)-2-(2-amino-6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine
J64468.03
J64468.03EA
82.9
EUR
InStock
J64468.03
6-Chloro-9-(β-D-ribofuranosyl)-9H-purin-2-amine 95%
2-Amino-6-chloropurine riboside
Purity: 95%
CAS No.: 2004-07-1
Molecular Formula: C10H12ClN5O4
Formula Weight: 301.69
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 165-167°
MDL No.: MFCD00005735