J65669.03
J65669.03EA
69.1
EUR
InStock
J65669.03
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine-3'-(2-cyanoethyl diisopropylphosphoramidite)
Purity: 98%
Molecular Formula: C48H54N7O8P
Formula Weight: 887.96
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00792622