J67368.AC
J67368.ACEA
111
EUR
InStock
J67368.AC
Or 95%, nanoparticules 30 nm OD1 (Abs. 526 nm)
Or
Purity: 95%
CAS No.: 7440-57-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00003436
Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM