Synonyms:
1,1,1,2,2,2-Hexaphenyldisilane
L04928.03
L04928.03EA
42.8
EUR
InStock
L04928.03
Hexaphényldisilane ≥98%
Hexaphényldisilane
Purity: 98%
CAS No.: 1450-23-3
Molecular Formula: (C6H5)3SiSi(C6H5)3
Formula Weight: 518.81
Melting Point: 368-370°
Beilstein Reference: 2024230