L06380.06
L06380.14
L06380.06EA
108
EUR
InStock
L06380.06
L06380.14
2-Nonyne-1-ol 96%
2-Nonyne-1-ol
Purity: 96%
CAS No.: 5921-73-3
Molecular Formula: CH3(CH2)5C≡CCH2OH
Formula Weight: 140.23
Refractive Index: 1.4560
Boiling Point: 109-111°/11mm
MDL No.: MFCD00041566