147020
147000
147040
147010
146040
147050
147052
147051
146000
146010
146020
146030
146050
LOVI147020EA
69.5
EUR
InStock
LOVI147020
LOVI147000
LOVI147040
LOVI147010
LOVI146040
LOVI147050
LOVI147052
LOVI147051
LOVI146000
LOVI146010
LOVI146020
LOVI146030
LOVI146050
Test kits, chlorine (DPD), CHECKIT®
Kits de tests environnementaux
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Enables the use of large path lengths
- The mirror optics makes use of the view through the entire length of the cell
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Information de livraison: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.