Test kits, chlorine (DPD), CHECKIT®
Fournisseur: Lovibond
147020 147000 147040 147010 146040 147050 147052 147051 146000 146010 146020 146030 146050
LOVI147020EA 69.5 EUR InStock
LOVI147020 LOVI147000 LOVI147040 LOVI147010 LOVI146040 LOVI147050 LOVI147052 LOVI147051 LOVI146000 LOVI146010 LOVI146020 LOVI146030 LOVI146050
Test kits, chlorine (DPD), CHECKIT®
Kits de tests environnementaux
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Information de livraison: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité