DNA transfection reagent, Glial-Mag
Fournisseur: OZ BIOSCIENCES
GL00250 GL00500 KGL0250
OZBIGL00250EA 238.56 EUR InStock
OZBIGL00250 OZBIGL00500 OZBIKGL0250
DNA transfection reagent, Glial-Mag
Réactifs de transfection
Glial-Mag is designed to reach optimal transfection efficiency with microglial cell lines and primary cells. Glial-Mag kit is the association of a specific magnetic nanoparticles formulation (Glial-Mag reagent), issued from our Magnetofection™ technology and a booster (Glial-Boost) designed to enhance transfection efficiency.
  • Ready to use
  • Highly efficient with microglial cell lines and primary cells
  • Low nucleic acid amount minimises toxicity
  • High level of nucleic acid compaction
  • Easy and straightforward protocol
  • Compatible with any culture medium
Magnetofection™ is a highly efficient gene delivery method that exploits magnetic force exerted upon gene vectors associated with magnetic particles to drive the nucleic acids towards and into the target cells. The nucleic acid dose gets concentrated onto the cells within a few minutes and 100% of cells get in contact with a significant vector dose.
Suitable for transfection of microglial cells line BV2, N9, N13, HMO6, MG-5, SIM-A9, primary microglia and transient and stable transfection.
Store at –20 °C.
Information de livraison: Shipped at room temperature.
Attention: For research purposes only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité