NIST-1764A
PROONIST-1764AEA
2450
EUR
InStock
PROONIST-1764A
Low alloy steel standard
Étalons
Étalons et alliages de métaux
This reference material is intended for use in the evaluation of chemical and instrumental analysis and in calibration of those methods.
A unit consists of a disk approximately 34 mm in diameter and 19 mm thick.
Certifications: Traceable to NIST and SRM.