NIST-3167A
PROONIST-3167AEA
1700
EUR
InStock
PROONIST-3167A
Yttrium standard solution, NIST-3167A
Étalons
Étalons cationiques
Standards d'yttrium
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of yttrium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically from high-purity yttrium (Ⅲ) oxide to contain a known mass fraction of yttrium.
Certifications: Traceable to NIST and SRM numbers.