32417
REST32417EA
58.2
EUR
InStock
REST32417
Organochlorine pesticide system evaluation mix (2 components)
Étalons
Étalons organiques
Contains two components, those are 4,4'-DDT and endrin.
- Designed for daily assessment of system performance
- Reveals active sites in the injection port and/or GC column
Prepared in MTBE—low expansion volume helps minimise backflash.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.