05855
05854
STMC05855EA
389
EUR
InStock
STMC05855
STMC05854
mFreSR™ Cryopreservation Medium
Milieux de cryoconservation cellulaire
Serum-free cryopreservation medium for human ES and iPS cells.
- Easy to use
- Serum-free formulation, optimised for use with TeSR™ maintenance media
- High thawing efficiencies
- Pre-screened components to ensure batch-to-batch consistency
mFreSR™ is a serum-free cryopreservation medium designed for the cryopreservation of human embryonic and induced pluripotent stem cells (ES and iPS cells). mFreSR™ contains DMSO and is complete and ready-to-use. Together with mTeSR™1 or mTeSR™ Plus, mFreSR™ eliminates the use of feeders and serum. Human ES and iPS cells cryopreserved in mFreSR™ have thawing efficiencies higher than reported conventional thawing methods using serum.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').