Wheat Germ Agglutinin (WGA), iFluor™ 647
Fournisseur: STEMCELL Technologies
100-0818
STMC100-0818EA 335 EUR InStock
STMC100-0818
Wheat Germ Agglutinin (WGA), iFluor™ 647
Protéines et Peptides
Fluorescent dye conjugates for staining cell membranes of gram-positive bacteria, mammalian cells, and yeast bud scars - 1000 µg.
Wheat germ agglutinins (WGA) are lectins or carbohydrate-binding proteins that have a high affinity for N-acetyl-D-glucosamine and sialic acid present in the cell membranes. Their iFluor™ conjugates are used to stain cell membranes and fibrotic scar tissue for fluorescence imaging and analysis.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité