Magnetic stirrers, MIXdrive 1 XL
Fournisseur: 2MAG
90600 90600_CH 90600_UK 40601
TMAG90600EA 1279 EUR InStock
TMAG90600 TMAG90600_CH TMAG90600_UK TMAG40601
Magnetic stirrers, MIXdrive 1 XL
Agitateurs Agitateurs magnétiques
Ultra-flat magnetic stirrers with stainless steel housing. Operating temperatures up to 100 °C.
  • Easy to clean and firm
  • Suitable for use with highly viscous media
  • Very strong magnetic coupling by SmCo magnets, for magnetic power transmission over extended distances, for thick vessel bottoms
Equipped as standard with mixWATCH ensuring a very high process safety, e.g. when using closed non-glass vessels or for unattended overnight operation. In case of the magnetic coupling’s separation between the magnetic stirrer and the stirring bar during the acceleration phase, the magnetic stirrer will be stopped, the stirring bar centred and stirring resumed.
IP protection class: IP 66
Certifications: IP protection class: IP 66
Informations de commande: Order MIXcontrol XL control unit separately.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité