Affinity chromatography columns, TSKgel®
Fournisseur: TOSOH Bioscience
0008645 0008647 0013066 0014440 0014449 0014451 0014455 0014456 0020022 0013125
TOYO0008645EA 1880 EUR InStock
TOYO0008645 TOYO0008647 TOYO0013066 TOYO0014440 TOYO0014449 TOYO0014451 TOYO0014455 TOYO0014456 TOYO0020022 TOYO0013125
Affinity chromatography columns, TSKgel®
Colonnes pour chromatographie
High efficiency resin based affinity chromatography columns will separate or purify many enzymes and other proteins with high level of specificity and selectivity.
  • Rigid polymeric base resin with high size exclusion limit (>5×10⁶ Da) and small particle size
  • Stable affinity ligands that enhances the access of large proteins to affinity ligands
  • High efficiency for analytical (10 µm) and semi-preparative (13 µm) affinity applications
  • Wide pH range (2 to 12) of the base resin, enabling robust cleaning options
  • Long lifetime, solvent compatibility, autoclavable, application flexibility, scalability from lab to commercial production
  • Eliminates undesirable interactions with column hardware
It consists of two group specific stationary phases such as TSKgel Boronate-5PW and TSKgel Chelate-5PW as well as one activated packing material called TSKgel Tresyl-5PW which can be readily scaled up for preparative separations.
Affinity columns are offered in glass and stainless steel. Stainless steel (SS) or Pyrex frits are embedded in the body of the column end-fittings of metal and glass columns, respectively. The nominal frit size for SS columns is engraved in the end-fittings. Pyrex frits in the glass columns have a 10 µm nominal pore size.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité