Synonyms:
2-Hydroxymethyltetrahydropyran, 2-(Hydroxymethyl)tetrahydro-2H-pyran
L17417.06
L17417.14
L17417.06EA
40.2
EUR
InStock
L17417.06
L17417.14
Tetrahydropyran-2-methanol ≥94%
Tetrahydropyran-2-methanol
Purity: 94%
CAS No.: 100-72-1
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 93°(199°F)
Refractive Index: 1.4580
Melting Point: -70°
Boiling Point: 186-189°
Beilstein Reference: 102998
MDL No.: MFCD00006624