Anti-TMEM16A Mouse Monoclonal Antibody (CF647) [clone: DOG1.1]
Fournisseur: Biotium
BNC470725-500 BNC470725-100
BNC470725-500EA 560 EUR InStock
BNC470725-500 BNC470725-100
Anti-TMEM16A Mouse Monoclonal Antibody (CF647) [clone: DOG1.1]
Anticorps
Expression of DOG-1 protein is elevated in the gastrointestinal stromal tumors (GISTs), c-kit signaling-driven mesenchymal tumors of the GI tract. DOG-1 is rarely expressed in other soft tissue tumors, which, due to appearance, may be difficult to diagnose. Immunoreactivity for DOG-1 has been reported in 97.8 percent of scorable GISTs, including all c-kit negative GISTs. Overexpression of DOG-1 has been suggested to aid in the identification of GISTs, including Platelet-Derived Growth Factor Receptor Alpha mutants that fail to express c-kit antigen. The overall sensitivity of DOG1 and c-kit in GISTs is nearly identical: 94.4% vs. 94.7%.
Type: Primary
Antigen: TMEM16A
Clonality: Monoclonal
Clone: DOG1.1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité