Synonyms:
2,2,2-Trifluoroethyl p-toluenesulphonate, 2,2,2-Trifluoroethyl 4-methylbenzenesulphonate
A17871.14
A17871.22
A17871.14EA
56.4
EUR
InStock
A17871.14
A17871.22
2,2,2-Trifluoroethyl toluene-4-sulphonate ≥98%
2,2,2-Trifluoroethyl toluene-4-sulphonate
Purity: 98+%
CAS No.: 433-06-7
Molecular Formula: C9H9F3O3S
Formula Weight: 254.23
Flash Point: 110°(230°F)
Melting Point: 37-42°
Boiling Point: 87-92°/0.1mm
Beilstein Reference: 2699394
MDL No.: MFCD00000443