Accredited Thermometer Recertification & Calibration Services
Accredited Thermometer Recertification & Calibration Services
VWRCATALYST offers recertification, calibration, and repair services for all makes and models of thermometers.
To order thermometer recertification and calibration, please complete all the information on the quote form and then carefully pack your thermometer along with the current calibration certificate.
  • Complete visual inspection for any physical damage. If possible the damage will be repaired.
  • Recertification from 1 Point to 15 Points.
  • Thermometer readings are compared against NIST standards in high precision calibration baths using the comparison method at determined test temperatures.
  • Each temperature is recorded to 1/10th of a scale division.
  • A calibration report is generated detailing the pertinent information including the correction factors for each test point.
  • All calibration information is entered into a data storage system.
  • All certification procedures are performed in accordance with NIST and ASTM guidelines, which meet with requirements set by regulatory agencies.
VWR Thermometer Calibration Sample Certification
Thermometer Recertification and Calibration Process