Final fill for recombinant proteins
Progressing from the drug substance to the final injectable drug product requires final packaging vials and stoppers that meet a number of quality requirements.
To meet these requirements, products must be prepared in an ISO Class 5 environment and meet USP71, USP85 and USP758 requirements to be sterile, pyrogen-free and particulate-free. They must also be validated for use in storage of frozen or lyophilized drug product and for compatibility with the protein and excipients in final formulation.
We also offer a range of excipients that are used in drug product formulation — including adjuvants, stabilizers, preservatives and bulking agents — and production water processed and meeting testing requirements in accordance with USP Water-for- Injection (WFI) regulations used in drug product manufacturing and as a diluent.
Explore solutions for final fill of recombinant proteins
Explore product choices for final fill for recombinant proteins
Ready to Get Started?
  • Contact us
  • Literature
  • Read case studies
  • Watch videos
Ready to Maximize Production Efficiency?
Our Bioprocessing Solutions team can help you streamline your biomanufacturing processes!
Get Started