3D PlatesFeatured Solutions![]() Corning Spheroid Microplate is ideal for generating and analysing 3D spheroids in the same microplate. Imaging plates with high optical quality are ideal for performing high content cell-based assays using imaging systems. Spheroid platesImaging plates New Elplasia plates ![]() The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids. Additional Resources |