02EFC06191
02EFC06191EA
3610
EUR
InStock
02EFC06191
Kanamycine sulfate
Kanamycine sulfate
Kanamycin sulfate is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus. It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene.
- Antibiotic
- Inhibitor of protein synthesis
Attention: Research or further manufacturing use only, not for direct human use.