Filtered through a 0.2 µm filter.
0323-07EA
1052.01
EUR
InStock
0323-07
Acide chlorhydrique 25%, Réactif Biotech, J.T.Baker®
Acide chlorhydrique
Made from USP purified water and hydrochloric acid, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.