04516-1L
04516-1LEA
108
EUR
InStock
04516-1L
Propanol-2 ≥99.9% (par GC), TraceSELECT™ pour analyse de la spéciation des métaux, Riedel-de Haën™
Propanol-2
High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.