Organic reference standard, alpha-Terpineol, analytical standard, Supelco®
Fournisseur: Merck
04899-250MG
04899-250MGEA 76.2 EUR InStock
04899-250MG
Organic reference standard, alpha-Terpineol, analytical standard, Supelco®
Étalons Étalons organiques
α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil.
  • Used in the synthesis of α-terpinyl esters of acetic acid and acetic anhydride via lipase-mediated esterification
The reaction rate constant of α-terpineol with OH radical and ozone was found to be (1,9±0,5)×10⁻¹⁰cm³ molecule⁻¹s⁻¹ and (3,0±0,2)×10⁻¹⁶ cm³ molecule⁻¹s⁻¹, respectively.
Helps in identification of α-terpineol in tequila by solid phase microextraction (SPME) and capillary gas chromatography-mass spectrometry (GC-MS), and in virgin olive oil samples by headspace SPME followed by GC-MS equipped with flame ionisation detector (FID).
utilised as a reference standard for the analysis of α-terpineol as trace volatile compound in wine by solid-phase extraction (SPE) followed by GC-MS and in essential oils and related compounds from aromatic plants by SPME and GC-MS.
Attention: Depending on the ambient or transport temperature the aggregation state can vary.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a3e045a1770a62e4bd3a02e033d8fb34619647&vid=f2a3e045a1770a62e4bd3a02e033d8fb34619647&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité