Organic reference standard, Hexacosanoic acid, analytical standard, Supelco®
Fournisseur: Merck
06922-10MG
06922-10MGEA 68.7 EUR InStock
06922-10MG
Organic reference standard, Hexacosanoic acid, analytical standard, Supelco®
Étalons Étalons organiques
Hexacosanoic acid is classified under the very long chain fatty acids (VLCFAs) group of biologically important compounds.
  • Identity confirmed by LC-MS method
Used as a reference standard for the determination of hexacosanoic acid in human serum and plasma samples for the screening of peroxisomal disorders by gas chromatography-mass spectrometry (GC-MS) operating under selected-ion monitoring (SIM) mode, in biological samples by GC-MS, in microalgal lipids by solid-phase extraction (SPE) followed by GC-MS analysis as a naphthoxyethyl derivative by high performance liquid chromatography (HPLC) coupled with fluorometric detection.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b17b5a3f8e29c89b5e793799c7aa45dc5ffc5fce&vid=b17b5a3f8e29c89b5e793799c7aa45dc5ffc5fce&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité