Phytochemical standard, alpha-Hederin, analytical standard, Supelco®
Fournisseur: Merck
07512-10MG
07512-10MGEA 237 EUR InStock
07512-10MG
Phytochemical standard, alpha-Hederin, analytical standard, Supelco®
Étalons Étalons organiques
α-Hederin is a water-soluble pentacyclic triterpene compound present in the seeds of Nigella sativa.
Utilised internal standard for the determination of glycyrrhizin and its major metabolite glycyrrhetic acid in human plasma by liquid chromatography/tandem mass spectrometry (LC−MS/MS) and also used as an external standard for quinovic acid glycosides assay by high performance liquid chromatography-photodiode array detection (HPLC-PDA) as well as ultra-performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UPLC/Q-TOF-MS).
Used as a reference standard for the identification of α-hederin in the leaves of common ivy (Hedera helix) by liquid chromatography-electrospray ionisation tandem mass spectrometry (LC-EI/MS-MS).
Conditionnement: Bottomless glass bottle contents are inside inserted with fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1fea8341e8a0d6b05f3e0759d12e6576f5ded64&vid=f1fea8341e8a0d6b05f3e0759d12e6576f5ded64&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité