1.00490.9025
1.00490.9025EA
0
EUR
InStock
1.00490.9025
Parteck® ODT, EMPROVE® ESSENTIAL, SAFC®
Parteck®
Parteck® ODT is a superior directly compressible excipient for fast disintegrating oral preparations. These solid dosage forms rapidly disintegrate on contact with saliva, thus eliminating the need for water and the difficulties some consumers have with swallowing tablets. This fast disintegration is a prerequisite for rapid dissolution of the active ingredients.