Acide chlorhydrique 0,1 mol/l (0,1 N), Titripur® Reag. USP, Reag. Ph. Eur. Volumetric, Supelco®
Fournisseur: Merck
Attention
Synonyms: Hydrochloric acid solution, Muriatic acid
MQ100
1.09060.1000 1.09060.5000 1.09060.9010 1.09060.9025 1.09060.4000 1.09060.1003 1.09060.4003 1.09060.9013
1.09060.1000EA 16.56 EUR InStock
1.09060.1000 1.09060.5000 1.09060.9010 1.09060.9025 1.09060.4000 1.09060.1003 1.09060.4003 1.09060.9013
Acide chlorhydrique 0,1 mol/l (0,1 N), Titripur® Reag. USP, Reag. Ph. Eur. Volumetric, Supelco®
Acide chlorhydrique
Merck have introduced into their Titripur® Titrants range packs with additional technology - Supelco® SmartTitrants use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. Mettler Toledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.
Formule: HCl
MW: 36.46 g/mol
Densité: 1.004 g/cm³ (25 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011324
Numéro CAS: 7647-01-0
EINECS: 231-595-7
ONU: 1789
ADR: 8 II
Commander maintenant
Résultats des tests de spécification
Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité