M-Clarity™ quality level = MQ200
1.09639.2500
1.09639.2500EA
152
EUR
InStock
1.09639.2500
(S)-(-)-Éthyle lactate, EMPLURA®
(S)-(-)-Éthyle lactate
Ethyl lactate is a safer and more sustainable alternative to ethyl acetate and acetone. It is an ester of natural L-lactic acid, which is produced by fermentation of sugar.
- Increased user safety due to less toxicity (non-carcinogenic)
- No waste due to 100% biodegradability
- Non-corrosive in contact with metals