M-Clarity™ quality level = MQ100
1.17048.1000
1.17048.1000EA
69
EUR
InStock
1.17048.1000
Acide sulfurique 95-97% pour la détermination de la DCO, Supelco®
Acide sulfurique
COD analysis according to e.g.
• ISO 6060
• ISO 15705
• APHA 5220 B
• APHA 5220 C
• EPA 410.2
• EPA 410.3
• DIN 38409 H41
• DIN 38409 H43
• All other COD norms
• Merck’s new COD Cell Tests for seawater / high chloride contents (1.17058.0001 and 1.17059.0001)