L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
DNA transfection reagents for protein production, FectoPRO®
Fournisseur: Polyplus-transfection
101000014 101000007 101000019
101000014.EA 542 EUR InStock
101000014. 101000007. 101000019.
DNA transfection reagents for protein production, FectoPRO®
Réactifs de transfection
The FectoPRO® Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various serum-free media, using low DNA amount (<1 μg/ml of cell culture).
  • Very high protein and antibody production yields
  • Cost-effective transient gene expression using lower plasmid DNA amount
  • Compatible with various mammalian expression media and cell systems
  • Compliant with biomanufacturing guidelines
  • Reproducible production yields
The FectoPRO®-mediated transfection process is easily scalable from a few millilitres to several litres of cell culture, ensuring robust reproducible protein production. Increased productivity and low DNA amount requirement make FectoPRO® a cost-effective solution for bioproduction processes.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number.
New product part numbers have symbol “New”. For duplicated products with symbol “telephone” please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
FectoPRO® is guaranteed free of animal-origin components, and is chemically defined. Systematic lot management and release testing is performed for each lot produced. Furthermore FectoPRO® transfection kit undergoes advanced quality controls: protein productivity, cell viability, and complete sterility.
Information de livraison: Each kit contains equal parts FectoPRO™ Reagent and FectoPRO™ Booster. FectoPRO™ Booster is also available separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..