10174.FI
10174.FIEA
0
EUR
InStock
10174.FI
Europium ≥99.9% (REO, base d'oxyde de terre rare), en feuilles, Thickness: 1.0 mm (0.04 in)
Europium
Purity: 99.9%
CAS No.: 7440-53-1
Physical Form: ≈3.3g/25x25mm, Packaged in mylar under argon
Odour: Odourless
UN No.: 3208
MDL No.: MFCD00010992