Aminobenzoate Potassium, Reference Standard
Fournisseur: USP
1019756
1019756.EA 245 EUR InStock
1019756.
Aminobenzoate Potassium, Reference Standard
Étalons Étalons organiques
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Collapse All
Désignation Normes Type de conditionnement Cdt Numéro de catalogue VWR Quantité
Aminobenzoate Potassium USP reference standard Vial 200 mg 1019756.
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité