Accessoires pour batterie Versaflo™ TR-830
Fournisseur: 3M
TR-838
111-2547EA 24.3 EUR InStock
111-2547
Accessoires pour batterie Versaflo™ TR-830
Piles
Enables attachment of TR-830 batteries to TR-802N PAPR blower motors
  • Bright yellow touchpoints
  • to differentiate from
  • non-IS versions of
  • Versaflo Product Family
  • Battery features bumpers
  • to help absorb impact
  • Mechanical attachment
  • of battery to help prevent
  • battery removal and
  • spark risk during use
  • per IEC/UL 60079
The TR-838 Battery Attachment Tool is used to mechanically attach TR-830 batteries to the TR-802N Intrinsically Safe (IS) PAPR blower motor as required by the most current UL standard (8th edition UL 913) for Intrinsically Safe PAPRs.
Certifications: UL-certified with an intrinsically safe (IS) rating of
Division 1: IS Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4,
under the most current standard (UL 60079, 6th Edition, 2013)
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité