Cut resistant gloves, Showa 67NFW
Fournisseur: Showa
67NFW-10
112-5881EA 205 EUR InStock
112-5881
Cut resistant gloves, Showa 67NFW
Gants
This soft, cotton flannel liner with natural rubber coating and wrinkle finish provides superior cut, abrasion, snag, and puncture resistance.
  • Wrinkle finish provides outstanding wet and dry grip
  • Protects hands from chemicals and abrasion while remaining elastic and breathable
  • Impermeable for working in damp or greasy environments
  • Ideal replacement for leather and ordinary rubber-coated gloves
It is most suitable for mechanical and automotive applications, construction work, shipping and receiving, and wood handling.
Certifications: EN 388:2016 (3142X); Cat. II
Food contact approved
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité