General purpose gloves, Showa 7166R
Fournisseur: Showa
7166R-10
112-5892EA 12.4 EUR InStock
112-5892
General purpose gloves, Showa 7166R
Gants
The 7166 general purpose gloves feature an impermeable full nitrile coating that protects hands from a range of hazards, including abrasion, oils, and chemicals.
  • Reinforced safety cuff prevents liquid and debris from entering the glove
  • Soft cotton liner controls the temperature and wicks away sweat
  • Rough surface grip is ideal for wet or greasy environments and increased protection
Typical applications include: Chemical spray and treatment, cutting roses and thorns, public utility, washing and cleaning, drainage, piping, guttering and drain pipe, plumbing, water/sewage treatment.
Certifications: EN 388:2016 (4111X); Cat. II
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité