Pleated masks with ties, non sterile, HM4, PUREZERO®
Fournisseur: Halyard
CLN1341TB
112-6508EA 155 EUR InStock
112-6508
Pleated masks with ties, non sterile, HM4, PUREZERO®
Masques Masques pour salles propres
The HM4 Pleated Mask with Ties is a three-layer mask with long ties that provides high particle, bacterial, and viral filtration efficiency. This mask is for industrial and cleanroom use. The mask construction consists of low-linting materials with a SO SOFT inner layer, a polyester foam nose band for added comfort, and anti-fog performance.
  • Three-layer mask
  • Long ties for easy donning
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the task with PUREZERO® HM4 non sterile face masks, which are specifically designed to meet the stringent requirements of cleanroom environments. This mask features a unique bicomponent material (polyester and polyethylene). They are manufactured in facilities with raw materials and components traceable to the original supplier.
Conditionnement: Air pulled, double-bagged. 50 masks per bag, 10 bags per case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité