Cleanroom hoods, CleanMax®
Fournisseur: LAKELAND CLEANMAX APPAREL
CTL713CM
114-9543EA 528 EUR InStock
114-9543
Cleanroom hoods, CleanMax®
Vêtements à usage unique
Clean manufactured disposable apparel are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All apparel provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Conditionnement: 100/case, individually packed.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité