Cleanroom sleeves, CleanMax®
Fournisseur: LAKELAND CLEANMAX APPAREL
CTL850CMP-18
114-9544EA 285 EUR InStock
114-9544
Cleanroom sleeves, CleanMax®
Vêtements à usage unique
Clean manufactured nonsterile sleeves with bound seams. Clean manufactured in a silicone-free environment.
  • Fluid repellent; tested in accordance with ASTM D6978 for chemotherapy permeation
  • Open fold for easier donning
  • Resistant to blood, body fluid and viral penetration
  • Resistant to blood borne pathogens
  • IEST-RP-CC003 Category I particle cleanliness
  • Latex-free and silicone-free
CleanMax® cleanroom garments for controlled environments provide optimal comfort, quality, and protection. Ideally suited for use in ISO Class 4 - 8 cleanrooms and all controlled environments. CleanMax® garments are easy to don, comfortable to wear, and are backed by Lakeland's 30+ years as a manufacturer of disposable protective apparel.
Conditionnement: Packaged in pairs in a protective outer bag for ante areas.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité