VWR® Disposable Cleanroom Socks
Fournisseur: VWR Collection
114-9707EA 1180 EUR InStock
114-9707
VWR® Disposable Cleanroom Socks
Vêtements à usage unique
VWR® Disposable cleanroom socks are designed to provide all day comfort inside a cleanroom. They are made from materials that prevent any particle generation while in use.
  • Single-use disposable socks
  • Poly/Nylon blend
  • Comfortable for extended use
  • Cleanroom compatible
  • Annex 1 compliant
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments. Suitable for use in ISO 4 to 6 (Class 10 to 1000) cleanrooms. Also available in a launderable format (see part numbers VWRI113-9936 - VWRI113-9939).
Used disposable socks can be recycled using the Fabrics and Clothing Zero Waste Box™ from TerraCycle.
Conditionnement: 400 pairs per case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité