Lingettes pour salles blanches, stériles, Anticon® Sterile Gold
Fournisseur: CONTEC
492226-942
115-0439EA 147 EUR InStock
115-0439
Lingettes pour salles blanches, stériles, Anticon® Sterile Gold
Lingettes Lingettes pour salles propres Lingettes pour salles propres, stériles
100% no-run interlock knit polyester wipes with sealed edges and increased sorbency.
  • Validated sterile; ideal for use in aseptic environments
  • Sealed edges to reduce particle and fibre generation
  • Patented textile technology creates a polyester wipe with high sorbency
  • Laundered and packed in a Class 4 cleanroom, with laser cut sealed edge
  • High sorptive capacity decreases the number of wipes required
  • Excellent choice for wiping and absorbing spills in critical cleanroom environments
  • Packed in small quantities in easy tear bags to reduce waste
Anticon® Sterile Gold Wipes are a sterile version of Anticon® Gold HeavyWeight™ wipes. These wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The proprietary sealed edges reduce the amount of fibres released from the wipe. These wipes exhibit very low particle and fibre counts, helping to minimise contamination. Ideal for use in the most critical of cleanroom environments. Ideal for disinfectant and solvent application, and wiping to dry.
Recommended for ISO Class 5 environments.
Conditionnement: These wipes are conveniently packaged in easy tear bags containing only 20 wipes per package to eliminate wastage.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité